Anti-KCNJ10 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: KCNJ10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77741-490
- 77858-437
- Antigen symbol:
- KCNJ10
- KCNJ10
- Antigen synonyms:
- inwardly rectifying subfamily J member 10|ATP dependent inwardly rectifying potassium channel Kir4.1|ATP sensitive inward rectifier potassium channel 10|ATP-dependent inwardly rectifying potassium channel Kir4.1|ATP-sensitive inward rectifier potassium channel 10|BIRK10|Glial ATP dependent inwardly rectifying potassium channel KIR4.1|Inward rectifier K(+) channel Kir1.2|Inward rectifier K+ channel KIR1.2|Inwardly rectifying potassium channel Kir1.2|KCJ10_HUMAN|KCNJ 10|KCNJ13 PEN|KIR1.2|KIR4.1|Potassium channel|Potassium channel inwardly rectifying subfamily J member 10|Potassium inwardly rectifying channel subfamily J member 10|SESAME
- inwardly rectifying subfamily J member 10|ATP dependent inwardly rectifying potassium channel Kir4.1|ATP sensitive inward rectifier potassium channel 10|ATP-dependent inwardly rectifying potassium channel Kir4.1|ATP-sensitive inward rectifier potassium channel 10|BIRK10|Glial ATP dependent inwardly rectifying potassium channel KIR4.1|Inward rectifier K(+) channel Kir1.2|Inward rectifier K+ channel KIR1.2|Inwardly rectifying potassium channel Kir1.2|KCJ10_HUMAN|KCNJ 10|KCNJ13 PEN|KIR1.2|KIR4.1|Potassium channel|Potassium channel inwardly rectifying subfamily J member 10|Potassium inwardly rectifying channel subfamily J member 10|SESAME
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P78508
- P78508
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human ATP-sensitive inward rectifier potassium channel 10 protein (165-379AA)
- Recombinant Human ATP-sensitive inward rectifier potassium channel 10 protein (165-379AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together