Anti-ADGRA2 Rabbit Polyclonal Antibody
77737-167
New Product
- Antigen symbol:ADGRA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96PE1
- Antigen synonyms:DKFZp434C211|DKFZp434J0911|FLJ14390|G protein coupled receptor 124|G-protein coupled receptor 124|GP124_HUMAN|GPR 124|Gpr124|KIAA1531|TEM 5|TEM5|Tumor endothelial marker 5|Tumor endothelial marker 5 precursor
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:DKFZp434C211
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IF: 1:200 to 1:1000, ELISA:1:10000
Type:
Antigen: ADGRA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse