Anti-KCNJ9 Rabbit Polyclonal Antibody
Catalog # 77740-134
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:KCNJ9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q92806
- Antigen synonyms:G protein activated inward rectifier potassium channel 3|G protein coupled inward rectifier potassium channel|G protein-activated inward rectifier potassium channel 3|GIRK-3|GIRK3|Inward rectifier K(+) channel Kir3.3|Inwardly rectifier K(+) channel Kir3.3|Inwardly rectifier K+ channel KIR3.3|inwardly rectifying subfamily J member 9|IRK9_HUMAN|KIR3.3|Potassium channel|Potassium channel inwardly rectifying subfamily J member 9|Potassium inwardly rectifying channel subfamily J member 9|Potassium inwardly rectifying channel subfamily J9
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:G protein activated inward rectifier potassium channel 3
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: KCNJ9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat