Anti-ITIH4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ITIH4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-344
- 77858-336
- Antigen symbol:
- ITIH4
- ITIH4
- Antigen synonyms:
- 35 kDa inter-alpha-trypsin inhibitor heavy chain H4|gp120|H4P|IHRP|Inter-alpha-inhibitor heavy chain 4|Inter-alpha-trypsin inhibitor family heavy chain-related protein|Inter-alpha-trypsin inhibitor heavy chain H4|inter-alpha-trypsin inhibitor, heavy chain-like, 1|ITI heavy chain H4|ITI-HC4|ITIH4_HUMAN|ITIHL1|OTTHUMP00000197120|OTTHUMP00000197121|OTTHUMP00000213834|OTTHUMP00000213869|PK-120|PK120|Plasma kallikrein sensitive glycoprotein 120|PRO1851
- 35 kDa inter-alpha-trypsin inhibitor heavy chain H4|gp120|H4P|IHRP|Inter-alpha-inhibitor heavy chain 4|Inter-alpha-trypsin inhibitor family heavy chain-related protein|Inter-alpha-trypsin inhibitor heavy chain H4|inter-alpha-trypsin inhibitor, heavy chain-like, 1|ITI heavy chain H4|ITI-HC4|ITIH4_HUMAN|ITIHL1|OTTHUMP00000197120|OTTHUMP00000197121|OTTHUMP00000213834|OTTHUMP00000213869|PK-120|PK120|Plasma kallikrein sensitive glycoprotein 120|PRO1851
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14624
- Q14624
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Inter-alpha-trypsin inhibitor heavy chain H4 protein (689-930AA)
- Recombinant human Inter-alpha-trypsin inhibitor heavy chain H4 protein (689-930AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together