Anti-NAT1 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NAT1
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-944
- 77743-945
- Antigen symbol:
- NAT1
- NAT1
- Antigen synonyms:
- AAC1| ARY1_HUMAN| Arylamide acetylase 1| Arylamine N acetyltransferase 1| Arylamine N-acetyltransferase 1| mNAT| Monomorphic arylamine N acetyltransferase| Monomorphic arylamine N-acetyltransferase| N acetyltransferase type 1| N-acetyltransferase type 1| NAT 1| NAT-1| NATI
- AAC1| ARY1_HUMAN| Arylamide acetylase 1| Arylamine N acetyltransferase 1| Arylamine N-acetyltransferase 1| mNAT| Monomorphic arylamine N acetyltransferase| Monomorphic arylamine N-acetyltransferase| N acetyltransferase type 1| N-acetyltransferase type 1| NAT 1| NAT-1| NATI
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P18440
- P18440
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Arylamine N-acetyltransferase 1 protein (1-288AA)
- Arylamine N-acetyltransferase 1 protein (1-288AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together