Anti-NGDN Rabbit Polyclonal Antibody
77744-416
New Product
- Antigen symbol:NGDN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8NEJ9
- Antigen synonyms:C14orf120|Neuroguidin|Centromere accumulated nuclear protein 1|CANu1|EIF4E-binding protein
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:EIF4E-binding protein
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: NGDN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat