Anti-KCND1 Rabbit Polyclonal Antibody
Catalog # 77740-105
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:KCND1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NSA2
- Antigen synonyms:KCND1_HUMAN|Kv4.1|mShal|OTTHUMP00000025805|OTTHUMP00000025806|Potassium voltage gated channel Shal related subfamily member 1|Potassium voltage gated channel subfamily D member 1|Potassium voltage-gated channel subfamily D member 1|Shal type potassium channel|Voltage gated potassium channel Kv4.1|Voltage gated potassium channel subunit Kv4.1|Voltage-gated potassium channel subunit Kv4.1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Voltage-gated potassium channel subunit Kv4.1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA:1:20000
Type:
Antigen: KCND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse