Anti-IFIT5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IFIT5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-643
- 77857-852
- Antigen symbol:
- IFIT5
- IFIT5
- Antigen synonyms:
- FLJ53857|FLJ92678|IFIT-5|IFIT5_HUMAN|Interferon induced protein with tetratricopeptide repeats 5|Interferon-induced protein with tetratricopeptide repeats 5|Retinoic acid and interferon inducible 58 kDa protein|Retinoic acid and interferon inducible protein (58kD)|Retinoic acid- and interferon-inducible 58 kDa protein|RI58
- FLJ53857|FLJ92678|IFIT-5|IFIT5_HUMAN|Interferon induced protein with tetratricopeptide repeats 5|Interferon-induced protein with tetratricopeptide repeats 5|Retinoic acid and interferon inducible 58 kDa protein|Retinoic acid and interferon inducible protein (58kD)|Retinoic acid- and interferon-inducible 58 kDa protein|RI58
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13325
- Q13325
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interferon-induced protein with tetratricopeptide repeats 5 protein (107-320AA)
- Recombinant Human Interferon-induced protein with tetratricopeptide repeats 5 protein (107-320AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together