Anti-NDUFB1 Rabbit Polyclonal Antibody
77744-165
New Product
- Antigen symbol:NDUFB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O75438
- Antigen synonyms:CI-MNLL|CI-SGDH|complex I MNLL subunit|Complex I-MNLL|MNLL|NADH dehydrogenase (ubiquinone) 1 beta subcomplex|1|7kDa|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 1|NADH-ubiquinone oxidoreductase 1 beta subcomplex|NADH-ubiquinone oxidoreductase MNLL subunit|NADH:ubiquinone oxidoreductase subunit B1|NDUB1_HUMAN|NDUFB1
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:CI-MNLL
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: NDUFB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human