Anti-NPR3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NPR3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-858
- 77744-859
- Antigen symbol:
- NPR3
- NPR3
- Antigen synonyms:
- ANP C|ANP-C|ANPC|ANPR-C|ANPRC|ANPRC_HUMAN|Atrial natriuretic peptide C type receptor |Atrial natriuretic peptide clearance receptor|Atrial natriuretic peptide receptor 3|Atrial natriuretic peptide receptor type C|Atrionatriuretic peptide receptor C|C5orf23|FLJ14054|GUCY2B|MGC22189|Natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C)|Natriuretic peptide receptor C/guanylate cyclase C|NPR 3|NPR C|NPR-C|Npr3|NPRC
- ANP C|ANP-C|ANPC|ANPR-C|ANPRC|ANPRC_HUMAN|Atrial natriuretic peptide C type receptor |Atrial natriuretic peptide clearance receptor|Atrial natriuretic peptide receptor 3|Atrial natriuretic peptide receptor type C|Atrionatriuretic peptide receptor C|C5orf23|FLJ14054|GUCY2B|MGC22189|Natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C)|Natriuretic peptide receptor C/guanylate cyclase C|NPR 3|NPR C|NPR-C|Npr3|NPRC
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17342
- P17342
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Atrial natriuretic peptide receptor 3 protein (36-127AA)
- Atrial natriuretic peptide receptor 3 protein (36-127AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together