Anti-GABPB1 Rabbit Polyclonal Antibody
77737-819
New Product
- Antigen symbol:GABPB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q06547
- Antigen synonyms:BABPB2|E4TF1|E4TF1-47|E4TF1-53|E4TF1B|GA binding protein transcription factor beta subunit 1 transcript variant gamma 2|GA binding protein transcription factor|beta subunit 1|beta subunit 2|GA-binding protein subunit beta-1|GABP subunit beta-1|GABP subunit beta-2|GABP1_HUMAN|GABPB|GABPB-1|GABPB-2|GABPB1|GABPB2|NRF2B1|NRF2B2|Nuclear respiratory factor 2|Transcription factor E4TF1-47|Transcription factor E4TF1-53
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Transcription factor E4TF1-53
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: GABPB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat