Order Entry
Canada
ContactUsLinkComponent
Anti-MAP3K2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-MAP3K2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MAP3K2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77859-394
  • 77742-668
  • Antigen symbol:
  • MAP3K2
  • MAP3K2
  • Antigen synonyms:
  • M3K2_HUMAN|Map3k2|MAPK/ERK kinase kinase 2|MEK kinase 2|MEKK 2|MEKK2b|Mitogen activated protein kinase kinase kinase 2|Mitogen-activated protein kinase kinase kinase 2
  • M3K2_HUMAN|Map3k2|MAPK/ERK kinase kinase 2|MEK kinase 2|MEKK 2|MEKK2b|Mitogen activated protein kinase kinase kinase 2|Mitogen-activated protein kinase kinase kinase 2
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9Y2U5
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Mitogen-activated protein kinase kinase kinase 2 protein (357-619AA)
  • Recombinant Human Mitogen-activated protein kinase kinase kinase 2 protein (357-619AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together