Anti-EME1 Rabbit Polyclonal Antibody
Catalog # 77735-846
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:EME1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96AY2
- Antigen synonyms:6820428D13|Crossover junction endonuclease EME1|EC 3.1.22.-|Eme1 essential meiotic endonuclease 1 homolog 1 (S. pombe)|S. pombe|homolog of|1|EME1_HUMAN|essential meiotic endonuclease 1 homolog 1 (S pombe)|Essential Meiotic Endonuclease 1 Homolog 1|essential meiotic endonuclease 1 homolog 2|Essential meiotic endonuclease 1|FLJ31364|hMMS4|homolog of yeast EME1 endonuclease|MGC106543|MMS4|MMS4 homolog|MMS4L
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:MMS4L
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: EME1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat