Anti-MOCS2 Rabbit Polyclonal Antibody
Catalog # 77743-293
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MOCS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:O96033
- Antigen synonyms:MOCS2|MOCO1|Molybdopterin synthase sulfur carrier subunit|MOCO1-A|Molybdenum cofactor synthesis protein 2 small subunit|Molybdenum cofactor synthesis protein 2A|MOCS2A|Molybdopterin-synthase small subunit|Sulfur carrier protein MOCS2A
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Sulfur carrier protein MOCS2A
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MOCS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat