Anti-FPR2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB:1:1000 to 1:5000, IF:1:50 to 1:200
Type:
Antigen: FPR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77855-593
- 77737-487
- Antigen symbol:
- FPR2
- FPR2
- Antigen synonyms:
- ALXR|FMLP-R-I|FMLP-R-II|FMLP-related receptor I|FMLPX|Formyl peptide receptor 2|Formyl peptide receptor related|Formyl peptide receptor-like 1|FPR2_HUMAN|FPR2A|FPRH1|FPRH2|FPRL1|HM63|Lipoxin A4 receptor|LXA4 receptor|LXA4R|N-formyl peptide receptor 2|RFP
- ALXR|FMLP-R-I|FMLP-R-II|FMLP-related receptor I|FMLPX|Formyl peptide receptor 2|Formyl peptide receptor related|Formyl peptide receptor-like 1|FPR2_HUMAN|FPR2A|FPRH1|FPRH2|FPRL1|HM63|Lipoxin A4 receptor|LXA4 receptor|LXA4R|N-formyl peptide receptor 2|RFP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25090
- P25090
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human N-formyl peptide receptor 2 protein (307-351AA)
- Recombinant Human N-formyl peptide receptor 2 protein (307-351AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together