Anti-KLK11 Rabbit Polyclonal Antibody
Catalog # 77741-765
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:KLK11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UBX7
- Antigen synonyms:Hippostasin|hK11|Kallikrein 11|Kallikrein 11 isoform 1 preproprotein|Kallikrein 11 isoform 2 precursor|Kallikrein 11 precursor|Kallikrein related peptidase 11|Kallikrein-11 inactive chain 2|KLK 11|KLK11_HUMAN|MGC33060|Protease serine 20 trypsin like|Protease serine trypsin like|PRSS20|Serine protease 20|TLSP|Trypsin like protease|Trypsin-like protease
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Trypsin-like protease
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: KLK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat