Anti-MGST1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MGST1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-734
- 77743-105
- Antigen symbol:
- MGST1
- MGST1
- Antigen synonyms:
- Glutathione S transferase 12|GST12|MGST 1|MGST|MGST I|MGST1_HUMAN|Microsomal glutathione S-transferase 1|Microsomal GST 1|Microsomal GST-1|Microsomal GST-I
- Glutathione S transferase 12|GST12|MGST 1|MGST|MGST I|MGST1_HUMAN|Microsomal glutathione S-transferase 1|Microsomal GST 1|Microsomal GST-1|Microsomal GST-I
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P10620
- P10620
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Microsomal glutathione S-transferase 1 protein (63-96AA)
- Recombinant Human Microsomal glutathione S-transferase 1 protein (63-96AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together