Anti-OR5M3 Rabbit Polyclonal Antibody
77736-639
New Product
- Antigen symbol:OR5M3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:Q8NGP4
- Antigen synonyms:OR5M3|Olfactory receptor 5M3|Olfactory receptor OR11-191
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Olfactory receptor OR11-191
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB:1:500 to 1:2000, IF:1:200 to 1:1000, ELISA:1:5000
Type:
Antigen: OR5M3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human