Anti-NME6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NME6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-563
- 77744-562
- Antigen symbol:
- NME6
- NME6
- Antigen synonyms:
- Inhibitor of p53 induced apoptosis alpha|Inhibitor of p53-induced apoptosis-alpha|IPIA ALPHA|IPIA-alpha|NDK 6|NDK6_HUMAN|NDP kinase 6|NM23 H6|nm23-H6|NM23H6|Nucleoside diphosphate kinase 6|OTTHUMP00000164810|OTTHUMP00000210736|OTTHUMP00000210737|OTTHUMP00000210738|OTTHUMP00000210741|OTTHUMP00000210742|OTTHUMP00000210744|OTTHUMP00000210745
- Inhibitor of p53 induced apoptosis alpha|Inhibitor of p53-induced apoptosis-alpha|IPIA ALPHA|IPIA-alpha|NDK 6|NDK6_HUMAN|NDP kinase 6|NM23 H6|nm23-H6|NM23H6|Nucleoside diphosphate kinase 6|OTTHUMP00000164810|OTTHUMP00000210736|OTTHUMP00000210737|OTTHUMP00000210738|OTTHUMP00000210741|OTTHUMP00000210742|OTTHUMP00000210744|OTTHUMP00000210745
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75414
- O75414
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nucleoside diphosphate kinase 6 protein (1-186AA)
- Nucleoside diphosphate kinase 6 protein (1-186AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together