Order Entry
Canada
ContactUsLinkComponent
Anti-F2R Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-F2R Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-F2R Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77855-052
  • 77736-357
  • Antigen symbol:
  • F2R
  • F2R
  • Antigen synonyms:
  • CF2R|Coagulation factor II (thrombin) receptor |Coagulation factor II receptor|F2R|HTR|PAR 1|PAR-1|PAR1|PAR1_HUMAN|Protease activated receptor 1|Proteinase activated receptor 1|Proteinase-activated receptor 1|Thrombin receptor|TR
  • CF2R|Coagulation factor II (thrombin) receptor |Coagulation factor II receptor|F2R|HTR|PAR 1|PAR-1|PAR1|PAR1_HUMAN|Protease activated receptor 1|Proteinase activated receptor 1|Proteinase-activated receptor 1|Thrombin receptor|TR
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Gene ID:
  • P25116
  • P25116
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Proteinase-activated receptor 1 protein (42-102AA)
  • Recombinant Human Proteinase-activated receptor 1 protein (42-102AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95% Protein G purified
  • >95% Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together