Anti-SLC49A3 Rabbit Polyclonal Antibody
Catalog # 77743-082
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:SLC49A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q6UXD7
- Antigen synonyms:SLC49A3|Major facilitator superfamily domain-containing protein 7|mfsd7|MFSD7_HUMAN|MYL5|Myosin light polypeptide 5 regulatory protein
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:MFSD7
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SLC49A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat