Anti-FLI1 Rabbit Polyclonal Antibody
Catalog # 77737-274
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:FLI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q01543
- Antigen synonyms:SIC1|FLI1 proto oncogene|FLI1 EWS fusion gene|Friend leukemia virus integration 1|FLI 1|EWSR 2|Viral integration region SIC 1|ERGB transcription factor|homolog of|ETS transcription factor|mouse|Sarcoma breakpoint region 2 (EWSR2)|EWSR2|Ewing Sarcoma breakpoint region 2|Friend leukemia integration 1 transcription factor|Proto-oncogene Fli-1|Transcription factor ERGB|FLI 1 proto oncogene|FLI1_HUMAN|Friend leukemia integration 1 (FLI1) transcription factor|Fli-1 proto-oncogene
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:homolog of
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: FLI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat