Anti-GPR75 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GPR75
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-894
- 77856-378
- Antigen symbol:
- GPR75
- GPR75
- Antigen synonyms:
- G protein coupled receptor 75|GPR chr2|GPR75_HUMAN|GPRchr2|OTTHUMP00000159608|Probable G protein coupled receptor 75|Probable G-protein coupled receptor 75|WI 31133|WI31133
- G protein coupled receptor 75|GPR chr2|GPR75_HUMAN|GPRchr2|OTTHUMP00000159608|Probable G protein coupled receptor 75|Probable G-protein coupled receptor 75|WI 31133|WI31133
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95800
- O95800
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Probable G-protein coupled receptor 75 protein (372-540AA)
- Recombinant Human Probable G-protein coupled receptor 75 protein (372-540AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together