Anti-MCM4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MCM4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-910
- 77859-594
- Antigen symbol:
- MCM4
- MCM4
- Antigen synonyms:
- 19G|AI325074|AU045576|CDC 21|CDC 54|CDC21|CDC21 homolog|CDC21, S. pombe, homolog of|CDC54|Cell division cycle 21, S. pombe, homolog of|DNA replication licensing factor MCM 4|DNA replication licensing factor MCM4|hCdc 21|hCdc21|Homolog of S. pombe cell devision cycle 21|KIAA4003|mcdc21|MCM 4|MCM 4 minichromosome maintenance deficient 4|mcm4|MCM4 minichromosome maintenance deficient 4|MCM4 minichromosome maintenance deficient 4, mitotin (S. cerevisiae)|MCM4_HUMAN|Mcmd4|MGC33310|Minichromosome maintenance 4|Minichromosome maintenance complex component 4|Minichromosome maintenance deficient (S. cerevisiae) 4|Minichromosome maintenance deficient 4|Minichromosome maintenance deficient 4 homolog (S. cerevisiae)|Minichromosome maintenance, S. cerevisiae, homolog of, 4|mKIAA4003|NKCD|NKGCD|P1 CDC21|P1-CDC21|P1CDC21
- 19G|AI325074|AU045576|CDC 21|CDC 54|CDC21|CDC21 homolog|CDC21, S. pombe, homolog of|CDC54|Cell division cycle 21, S. pombe, homolog of|DNA replication licensing factor MCM 4|DNA replication licensing factor MCM4|hCdc 21|hCdc21|Homolog of S. pombe cell devision cycle 21|KIAA4003|mcdc21|MCM 4|MCM 4 minichromosome maintenance deficient 4|mcm4|MCM4 minichromosome maintenance deficient 4|MCM4 minichromosome maintenance deficient 4, mitotin (S. cerevisiae)|MCM4_HUMAN|Mcmd4|MGC33310|Minichromosome maintenance 4|Minichromosome maintenance complex component 4|Minichromosome maintenance deficient (S. cerevisiae) 4|Minichromosome maintenance deficient 4|Minichromosome maintenance deficient 4 homolog (S. cerevisiae)|Minichromosome maintenance, S. cerevisiae, homolog of, 4|mKIAA4003|NKCD|NKGCD|P1 CDC21|P1-CDC21|P1CDC21
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P33991
- P33991
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DNA replication licensing factor MCM4 protein (2-141AA)
- Recombinant Human DNA replication licensing factor MCM4 protein (2-141AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together