Anti-SRPK3 Rabbit Polyclonal Antibody
77737-357
New Product
- Antigen symbol:SRPK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UPE1
- Antigen synonyms:MSSK-1|MSSK1|Muscle specific serine kinase 1|Muscle-specific serine kinase 1|Serine/arginine rich protein specific kinase 3|Serine/arginine-rich protein-specific kinase 3|Serine/threonine protein kinase 23|Serine/threonine protein kinase SRPK3|Serine/threonine-protein kinase 23|SFRS protein kinase 3|SR-protein-specific kinase 3|Srpk3|SRPK3_HUMAN|SRSF protein kinase 3|STK 23|STK23
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:MSSK-1
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:10000
Type:
Antigen: SRPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat