Anti-NRAS Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: NRAS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77745-003
- 77745-004
- Antigen symbol:
- NRAS
- NRAS
- Antigen synonyms:
- ALPS4|AV095280|GTPase NRas|HRAS1|N ras|N ras protein part 4|Neuroblastoma RAS viral (v ras) oncogene homolog|NRAS|NRAS1|NS6|OTTHUMP00000013879|OTTMUSP00000023521|RASN_HUMAN|Transforming protein N Ras|Transforming protein N-Ras|v ras neuroblastoma RAS viral oncogene homolog
- ALPS4|AV095280|GTPase NRas|HRAS1|N ras|N ras protein part 4|Neuroblastoma RAS viral (v ras) oncogene homolog|NRAS|NRAS1|NS6|OTTHUMP00000013879|OTTMUSP00000023521|RASN_HUMAN|Transforming protein N Ras|Transforming protein N-Ras|v ras neuroblastoma RAS viral oncogene homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P01111
- P01111
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- GTPase NRas protein (1-186AA)
- GTPase NRas protein (1-186AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together