Anti-NCOA4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: NCOA4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-047
- 77744-046
- Antigen symbol:
- NCOA4
- NCOA4
- Antigen synonyms:
- 70 kDa androgen receptor activator|70 kDa androgen receptor coactivator|70 kDa AR activator|70 kDa AR-activator|Androgen receptor coactivator 70 kD|Androgen receptor coactivator 70 kDa protein|Androgen receptor-associated protein of 70 kDa|ARA 70|ARA70|DKFZp762E1112|ELE 1|ELE1|ELE1/ret TK|NCOA 4|NCoA-4|NCOA4|NCOA4_HUMAN|Nuclear receptor coactivator 4|Papillary thyroid carcinoma 3|PTC 3|PTC3|RET activating gene ELE1|Ret activating protein ELE1|Ret-activating protein ELE1|RFG
- 70 kDa androgen receptor activator|70 kDa androgen receptor coactivator|70 kDa AR activator|70 kDa AR-activator|Androgen receptor coactivator 70 kD|Androgen receptor coactivator 70 kDa protein|Androgen receptor-associated protein of 70 kDa|ARA 70|ARA70|DKFZp762E1112|ELE 1|ELE1|ELE1/ret TK|NCOA 4|NCoA-4|NCOA4|NCOA4_HUMAN|Nuclear receptor coactivator 4|Papillary thyroid carcinoma 3|PTC 3|PTC3|RET activating gene ELE1|Ret activating protein ELE1|Ret-activating protein ELE1|RFG
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13772
- Q13772
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Nuclear receptor coactivator 4 protein (1-280AA)
- Nuclear receptor coactivator 4 protein (1-280AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together