Anti-ME1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: ME1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-613
- 77742-937
- Antigen symbol:
- ME1
- ME1
- Antigen synonyms:
- Cytosolic malic enzyme 1|HUMNDME|Malate dehydrogenase|Malic enzyme 1|NADP(+) dependent|cytosolic|soluble|Malic enzyme|cytoplasmic|MAOX_HUMAN|MES|NADP dependent malic enzyme|NADP ME|NADP-dependent malic enzyme|NADP-ME|Pyruvic malic carboxylase
- Cytosolic malic enzyme 1|HUMNDME|Malate dehydrogenase|Malic enzyme 1|NADP(+) dependent|cytosolic|soluble|Malic enzyme|cytoplasmic|MAOX_HUMAN|MES|NADP dependent malic enzyme|NADP ME|NADP-dependent malic enzyme|NADP-ME|Pyruvic malic carboxylase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48163
- P48163
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human NADP-dependent malic enzyme protein (161-519AA)
- Recombinant Human NADP-dependent malic enzyme protein (161-519AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together