Order Entry
Canada
ContactUsLinkComponent
Anti-Fxn Rabbit Recombinant Polyclonal Antibody (Biotin)
Anti-Fxn Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-Fxn Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77737-745
  • 77855-666
  • Antigen symbol:
  • Fxn
  • Fxn
  • Antigen synonyms:
  • FrdaFrataxin|mitochondrial|EC 1.16.3.1) [Cleaved into: Frataxin intermediate form|Frataxin mature form]
  • FrdaFrataxin|mitochondrial|EC 1.16.3.1) [Cleaved into: Frataxin intermediate form|Frataxin mature form]
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O35943
  • O35943
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant mouse Frataxin, mitochondrial protein (78-207AA)
  • Recombinant mouse Frataxin, mitochondrial protein (78-207AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together