Anti-FFAR2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FFAR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-073
- 77855-308
- Antigen symbol:
- FFAR2
- FFAR2
- Antigen synonyms:
- Fatty acid receptor 2|FFA2R|FFAR2_HUMAN|free fatty acid activated receptor 2|Free fatty acid receptor 2|G protein coupled receptor 43|G protein-coupled receptor 43|G-protein coupled receptor 43|GPR 43|GPR43
- Fatty acid receptor 2|FFA2R|FFAR2_HUMAN|free fatty acid activated receptor 2|Free fatty acid receptor 2|G protein coupled receptor 43|G protein-coupled receptor 43|G-protein coupled receptor 43|GPR 43|GPR43
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15552
- O15552
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Free fatty acid receptor 2 protein (277-330AA)
- Recombinant Human Free fatty acid receptor 2 protein (277-330AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together