Anti-LTF Rabbit Recombinant polyclonal Antibody
New Product
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: LTF
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77742-448
- 77859-212
- Antigen symbol:
- LTF
- LTF
- Antigen synonyms:
- GIG12| Growth inhibiting protein 12| HLF2| Kaliocin 1| Lactoferrin| Lactoferroxin-C| Lactotransferrin| LF| Neutrophil lactoferrin| Talalactoferrin| TRFL_HUMAN
- GIG12|Growth inhibiting protein 12|HLF2|Kaliocin 1|Lactoferrin|Lactoferroxin-C|Lactotransferrin|LF|Neutrophil lactoferrin|Talalactoferrin|TRFL_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P02788
- P02788
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lactotransferrin protein (156-413AA)
- Recombinant Human Lactotransferrin protein (156-413AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together