11946
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77855-446
- 77737-250
- Antigen synonyms:
- BDPLT 6|BDPLT6|Cellular myosin heavy chain|Cellular myosin heavy chain type A|DFNA 17|DFNA17|EPSTS|FTNS|MGC104539|MHA|MYH 2A|MYH 9|MYH2A|MYH9|MYH9_HUMAN|MYHas8|MyHC 2A|MyHC IIa|MyHC2A|MyHCIIa|MYHSA 2|MYHSA2|Myosin 9|Myosin heavy chain 9|Myosin heavy chain 9 non muscle|Myosin heavy chain|Myosin heavy chain non muscle IIa|Myosin heavy chain nonmuscle IIa|Myosin heavy polypeptide 2|Myosin heavy polypeptide 9 non muscle|Myosin-9|Myosin9|NMHC II A|NMMHC A|NMMHC II a|NMMHC II-a|NMMHC IIA|NMMHC-A|NMMHC-IIA|NMMHCA|Non muscle myosin heavy chain A|Non muscle myosin heavy chain|Non muscle myosin heavy chain II A|Non muscle myosin heavy polypeptide 9|non-muscle IIa|Non-muscle myosin heavy chain A|Non-muscle myosin heavy chain IIa|Nonmuscle myosin heavy chain A|Nonmuscle myosin heavy chain II A|type A
- Gene ID:
- P35579
- Immunogen:
- Recombinant Human Myosin-9 protein (2-241AA)
- Form:
- Liquid
- Purification:
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
Specifications
Frequently Bought Together