Order Entry
Canada
ContactUsLinkComponent
Anti-GSPT1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-GSPT1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-GSPT1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77739-117
  • 77856-515
  • Antigen symbol:
  • GSPT1
  • GSPT1
  • Antigen synonyms:
  • eRF3a|ERF3AEukaryotic peptide chain release factor GTP-binding subunit ERF3A|G1 to S phase transition protein 1 homolog|Eukaryotic peptide chain release factor subunit 3a
  • ERF3AEukaryotic peptide chain release factor GTP-binding subunit ERF3A|Eukaryotic peptide chain release factor subunit 3a|eRF3a|G1 to S phase transition protein 1 homolog
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P15170
  • P15170
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Eukaryotic peptide chain release factor GTP-binding subunit ERF3A protein (6-142AA)
  • Recombinant Human Eukaryotic peptide chain release factor GTP-binding subunit ERF3A protein (6-142AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together