Anti-ADGRL2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: ADGRL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77859-105
- 77742-319
- Antigen symbol:
- ADGRL2
- ADGRL2
- Antigen synonyms:
- Adgrl2| Adhesion G protein-coupled receptor L2| AGRL2_HUMAN| Calcium-independent alpha-latrotoxin receptor 2| CIRL-2| CL2| KIAA0786| Latrophilin 1| Latrophilin 2| Latrophilin homolog 1| latrophilin homolog 2| Latrophilin-2| LEC1| Lectomedin-1| LPH2| LPHH1| LPHN2_HUMAN
- Adgrl2|Adhesion G protein-coupled receptor L2|AGRL2_HUMAN|Calcium-independent alpha-latrotoxin receptor 2|CIRL-2|CL2|KIAA0786|Latrophilin 1|Latrophilin 2|Latrophilin homolog 1|latrophilin homolog 2|Latrophilin-2|LEC1|Lectomedin-1|LPH2|LPHH1|LPHN2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95490
- O95490
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Adhesion G protein-coupled receptor L2 protein (400-480AA)
- Recombinant Human Adhesion G protein-coupled receptor L2 protein (400-480AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together