Anti-KHDRBS1 Rabbit Polyclonal Antibody
Catalog # 77741-562
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:KHDRBS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:Q07666
- Antigen synonyms:FLJ34027|GAP associated tyrosine phosphoprotein p62|GAP-associated tyrosine phosphoprotein p62|KH domain containing RNA binding signal transduction associated 1|KH domain-containing|KHDR1_HUMAN|Khdrbs1|p21 Ras GTPase activating protein associated p62|p21 Ras GTPase-activating protein-associated p62|p62|p68|RNA-binding|Sam68|signal transduction-associated protein 1|Src associated in mitosis 68 kDa protein|Src-associated in mitosis 68 kDa protein
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Src-associated in mitosis 68 kDa protein
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: KHDRBS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat