Anti-MED7 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: MED7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77859-638
- 77742-977
- Antigen symbol:
- MED7
- MED7
- Antigen synonyms:
- Activator recruited cofactor 34 kDa component|Activator-recruited cofactor 34 kDa component|ARC34|Cofactor required for Sp1 transcriptional activation subunit 9|Cofactor required for Sp1 transcriptional activation|subunit 9 (33kD)|subunit 9|33kDa|CRSP complex subunit 9|CRSP33|CRSP9|hMED7_HUMAN|Mediator complex subunit 7|Mediator of RNA polymerase II transcription subunit 7|MGC12284|RNA polymerase transcriptional regulation mediator subunit 7 homolog|Transcriptional coactivator CRSP33
- Activator recruited cofactor 34 kDa component|Activator-recruited cofactor 34 kDa component|ARC34|Cofactor required for Sp1 transcriptional activation subunit 9|Cofactor required for Sp1 transcriptional activation|subunit 9 (33kD)|subunit 9|33kDa|CRSP complex subunit 9|CRSP33|CRSP9|hMED7_HUMAN|Mediator complex subunit 7|Mediator of RNA polymerase II transcription subunit 7|MGC12284|RNA polymerase transcriptional regulation mediator subunit 7 homolog|Transcriptional coactivator CRSP33
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43513
- O43513
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mediator of RNA polymerase II transcription subunit 7 protein (1-233AA)
- Recombinant Human Mediator of RNA polymerase II transcription subunit 7 protein (1-233AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together