Anti-GIPC1 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
0
Type:
Antigen: GIPC1
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77856-002
- 77738-272
- Antigen symbol:
- GIPC1
- GIPC1
- Antigen synonyms:
- C19orf3|GIPC|RGS19IP1PDZ domain-containing protein GAIP C-terminus-interacting protein|RGS-GAIP-interacting protein|RGS19-interacting protein 1|Synectin|Tax interaction protein 2|TIP-2
- C19orf3| GIPC| RGS19IP1PDZ domain-containing protein GAIP C-terminus-interacting protein| RGS-GAIP-interacting protein| RGS19-interacting protein 1| Synectin| Tax interaction protein 2| TIP-2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14908
- O14908
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human PDZ domain-containing protein GIPC1 protein (7-74AA)
- Recombinant Human PDZ domain-containing protein GIPC1 protein (7-74AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together