Anti-LANCL1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LANCL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-835
- 77741-980
- Antigen symbol:
- LANCL1
- LANCL1
- Antigen synonyms:
- 40 kDa erythrocyte membrane protein| G protein-coupled receptor 69A| GPR69A| LanC (bacterial lantibiotic synthetase component C) like 1| LanC lantibiotic synthetase component C like 1 (bacterial)| LanC-like protein 1| LANC1_HUMAN| LANCL1| p40
- 40 kDa erythrocyte membrane protein|G protein-coupled receptor 69A|GPR69A|LanC (bacterial lantibiotic synthetase component C) like 1|LanC lantibiotic synthetase component C like 1 (bacterial)|LanC-like protein 1|LANC1_HUMAN|LANCL1|p40
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43813
- O43813
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human LanC-like protein 1 protein (50-150AA)
- Recombinant Human LanC-like protein 1 protein (50-150AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together