Anti-Krt18 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: Krt18
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
- Catalog No:
- 77858-737
- 77741-869
- Antigen symbol:
- Krt18
- Krt18
- Antigen synonyms:
- Kerd|Krt1-18|Keratin|type I cytoskeletal 18|Cytokeratin endo B|Keratin D|Cytokeratin-18|CK-18|Keratin-18|K18
- Kerd|Krt1-18|Keratin|type I cytoskeletal 18|Cytokeratin endo B|Keratin D|Cytokeratin-18|CK-18|Keratin-18|K18
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P05784
- P05784
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Mouse Keratin, type I cytoskeletal 18 protein (200-423AA)
- Recombinant Mouse Keratin, type I cytoskeletal 18 protein (200-423AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together