Anti-IVD Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IVD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-372
- 77858-355
- Antigen symbol:
- IVD
- IVD
- Antigen synonyms:
- ACAD2|FLJ12715|FLJ34849|Isovaleryl CoA dehydrogenase|Isovaleryl CoA dehydrogenase, mitochondrial|Isovaleryl Coenzyme A dehydrogenase|Isovaleryl-CoA dehydrogenase|IVD|IVD_HUMAN|mitochondrial
- Isovaleryl CoA dehydrogenase|mitochondrial|Isovaleryl CoA dehydrogenase, mitochondrial|IVD|FLJ12715|IVD_HUMAN|Isovaleryl-CoA dehydrogenase|ACAD2|Isovaleryl Coenzyme A dehydrogenase|FLJ34849
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P26440
- P26440
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Isovaleryl-CoA dehydrogenase, mitochondrial protein (139-241AA)
- Recombinant Human Isovaleryl-CoA dehydrogenase, mitochondrial protein (139-241AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together