Anti-FES Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: FES
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-292
- 77737-054
- Antigen symbol:
- FES
- Antigen synonyms:
- c-fes/fps protein|cFES|Feline sarcoma (Snyder Theilen) viral (v fes)/Fujinami avian sarcoma (PRCII) viral (v fps) oncogene homolog|feline sarcoma (Snyder-Theilen) viral (v-fes)/Fujinami|Feline sarcoma oncogene|Feline sarcoma/Fujinami avian sarcoma oncogene homolog|fes|FES proto oncogene|tyrosine kinase|FES_HUMAN|FPS|Oncogene FES|Oncogene FES feline sarcoma virus|P93c fes|Proto oncogene tyrosine protein kinase Fes/Fps|Proto-oncogene c-Fes|Proto-oncogene c-Fps|proto-oncogene tyrosine-protein kinase Fes/Fps|Tyrosine-protein kinase Fes/Fps
- c-fes/fps protein|cFES|Feline sarcoma (Snyder Theilen) viral (v fes)/Fujinami avian sarcoma (PRCII) viral (v fps) oncogene homolog|feline sarcoma (Snyder-Theilen) viral (v-fes)/Fujinami|Feline sarcoma oncogene|Feline sarcoma/Fujinami avian sarcoma oncogene homolog|fes|FES proto oncogene|tyrosine kinase|FES_HUMAN|FPS|Oncogene FES|Oncogene FES feline sarcoma virus|P93c fes|Proto oncogene tyrosine protein kinase Fes/Fps|Proto-oncogene c-Fes|Proto-oncogene c-Fps|proto-oncogene tyrosine-protein kinase Fes/Fps|Tyrosine-protein kinase Fes/Fps
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P07332
- P07332
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Tyrosine-protein kinase Fes/Fps protein (120-420AA)
- Recombinant Human Tyrosine-protein kinase Fes/Fps protein (120-420AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together