Anti-MTSS1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MTSS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77860-116
- 77743-627
- Antigen symbol:
- MTSS1
- MTSS1
- Antigen synonyms:
- 2310003N14Rik|BC024131|D130001D01Rik|DKFZp781P2223|FLJ44694|KIAA0429|metastasis suppressor 1|Metastasis suppressor protein 1|Metastasis suppressor YGL 1|Metastasis suppressor YGL-1|MGC37896|MIM|MIMA|MIMB|missing in metastasis|Missing in metastasis protein|mKIAA0429|MTSS 1|MTSS1_HUMAN
- 2310003N14Rik|BC024131|D130001D01Rik|DKFZp781P2223|FLJ44694|KIAA0429|metastasis suppressor 1|Metastasis suppressor protein 1|Metastasis suppressor YGL 1|Metastasis suppressor YGL-1|MGC37896|MIM|MIMA|MIMB|missing in metastasis|Missing in metastasis protein|mKIAA0429|MTSS 1|MTSS1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43312
- O43312
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Metastasis suppressor protein 1 protein (616-730AA)
- Recombinant human Metastasis suppressor protein 1 protein (616-730AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together