Anti-GPD1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: GPD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77856-299
- 77738-733
- Antigen symbol:
- GPD1
- GPD1
- Antigen synonyms:
- AI747587|cytoplasmic|EC 1.1.1.8|FLJ26652|G3PD|Gdc-1|Gdc1|Gdp1|Glycerol 3 phosphate dehydrogenase 1|Glycerol 3 phosphate dehydrogenase cytosolic|Glycerol 3 phosphate dehydrogenase soluble|Glycerol-3-phosphate dehydrogenase [NAD+]|Glycerol-3-phosphate dehydrogenase 1 (soluble)|Glycerol-3-phosphate dehydrogenase|soluble|Glycerphosphate dehydrogenase|GPD-C|Gpd1 protein|Gpd3|GPDA_HUMAN|Gpdc|GPDH|GPDH-C|Gpdhc|HTGTI|KIAA4010|MGC93453|MGPD|mKIAA4010
- AI747587|cytoplasmic|EC 1.1.1.8|FLJ26652|G3PD|Gdc-1|Gdc1|Gdp1|Glycerol 3 phosphate dehydrogenase 1|Glycerol 3 phosphate dehydrogenase cytosolic|Glycerol 3 phosphate dehydrogenase soluble|Glycerol-3-phosphate dehydrogenase [NAD+]|Glycerol-3-phosphate dehydrogenase 1 (soluble)|Glycerol-3-phosphate dehydrogenase|soluble|Glycerphosphate dehydrogenase|GPD-C|Gpd1 protein|Gpd3|GPDA_HUMAN|Gpdc|GPDH|GPDH-C|Gpdhc|HTGTI|KIAA4010|MGC93453|MGPD|mKIAA4010
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21695
- P21695
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Glycerol-3-phosphate dehydrogenase [NAD(+)], cytoplasmic protein (1-349AA)
- Recombinant Human Glycerol-3-phosphate dehydrogenase [NAD(+)], cytoplasmic protein (1-349AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together