Anti-ELOVL4 Rabbit Polyclonal Antibody
77735-836
New Product
- Antigen symbol:ELOVL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9GZR5
- Antigen synonyms:3-keto acyl-CoA synthase ADMD|Cancer/testis antigen 118|CT118|Elongation of very long chain fatty acids (FEN1/Elo2 SUR4/Elo3 yeast) like 4|Elongation of very long chain fatty acids like 4|Elongation of very long chain fatty acids protein 4|ELOV L4|ELOV4_HUMAN|ELOVL 4|FLJ17667|FLJ92876|Stargardt disease 3|Stargardt disease 3 autosomal dominant|STGD 2|STGD 3|STGD2|STGD3
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:STGD3
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human