Anti-LAIR2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: LAIR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77858-810
- 77741-951
- Antigen symbol:
- LAIR2
- LAIR2
- Antigen synonyms:
- CD306|CD306 antigen|LAIR 2|LAIR-2|LAIR2_HUMAN|Leukocyte associated Ig like receptor 2|Leukocyte associated immunoglobulin like receptor 2|Leukocyte-associated immunoglobulin-like receptor 2|MGC71634|OTTHUMP00000068389|OTTHUMP00000068390
- LAIR2_HUMAN|MGC71634|LAIR-2|Leukocyte associated Ig like receptor 2|LAIR 2|OTTHUMP00000068390|CD306|OTTHUMP00000068389|Leukocyte associated immunoglobulin like receptor 2|CD306 antigen|Leukocyte-associated immunoglobulin-like receptor 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ISS4
- Q6ISS4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leukocyte-associated immunoglobulin-like receptor 2 protein (22-152AA)
- Recombinant Human Leukocyte-associated immunoglobulin-like receptor 2 protein (22-152AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together