Anti-FOXA3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:500 to 1:1000
Type:
Antigen: FOXA3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-397
- 77855-534
- Antigen symbol:
- FOXA3
- FOXA3
- Antigen synonyms:
- FKHH3|Fork head-related protein FKH H3|forkhead box A3|Forkhead box protein A3|FOXA3_HUMAN|hepatic nuclear factor-3-beta|hepatocyte nuclear factor 3|hepatocyte nuclear factor 3 gamma|Hepatocyte nuclear factor 3-gamma|HNF-3-gamma|HNF-3G|HNF3B|HNF3G|TCF-3G|TCF3G|Transcription factor 3G
- FKHH3|Fork head-related protein FKH H3|forkhead box A3|Forkhead box protein A3|FOXA3_HUMAN|hepatic nuclear factor-3-beta|hepatocyte nuclear factor 3|hepatocyte nuclear factor 3 gamma|Hepatocyte nuclear factor 3-gamma|HNF-3-gamma|HNF-3G|HNF3B|HNF3G|TCF-3G|TCF3G|Transcription factor 3G
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55318
- P55318
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Hepatocyte nuclear factor 3-gamma protein (220-336AA)
- Recombinant Human Hepatocyte nuclear factor 3-gamma protein (220-336AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together