Anti-LAMA2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: LAMA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77858-813
- 77741-954
- Antigen symbol:
- LAMA2
- LAMA2
- Antigen synonyms:
- LAMA 2|LAMA2_HUMAN|Laminin alpha 2 (merosin congenital muscular dystrophy)|Laminin alpha 2|Laminin alpha 2 chain|Laminin alpha 2 subunit|Laminin M|Laminin M chain|Laminin subunit alpha-2|Laminin-12 subunit alpha|Laminin-2 subunit alpha|Laminin-4 subunit alpha|LAMM|Merosin heavy chain
- LAMA 2|LAMA2_HUMAN|Laminin alpha 2 (merosin congenital muscular dystrophy)|Laminin alpha 2|Laminin alpha 2 chain|Laminin alpha 2 subunit|Laminin M|Laminin M chain|Laminin subunit alpha-2|Laminin-12 subunit alpha|Laminin-2 subunit alpha|Laminin-4 subunit alpha|LAMM|Merosin heavy chain
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P24043
- P24043
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Laminin subunit alpha-2 protein (974-1162AA)
- Recombinant Human Laminin subunit alpha-2 protein (974-1162AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together