Anti-NUP153 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NUP153
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77745-208
- 77745-209
- Antigen symbol:
- NUP153
- NUP153
- Antigen synonyms:
- 153 kDa nucleoporin|HN153|NU153_HUMAN|Nuclear pore complex protein hNuclear pore complex protein Nucleoporin 153kDa|Nucleoporin Nup 153|Nup153
- 153 kDa nucleoporin|HN153|NU153_HUMAN|Nuclear pore complex protein hNuclear pore complex protein Nucleoporin 153kDa|Nucleoporin Nup 153|Nup153
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49790
- P49790
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nuclear pore complex protein Nup153 protein (657-880AA)
- Nuclear pore complex protein Nup153 protein (657-880AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together