Anti-NME5 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NME5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-553
- 77744-554
- Antigen symbol:
- NME5
- NME5
- Antigen synonyms:
- Inhibitor of p53 induced apoptosis beta|Inhibitor of p53-induced apoptosis-beta|IPIA beta|IPIA-beta|NDK-H 5|NDK5_HUMAN|NDP kinase homolog 5|NM23 H5|nm23-H5|Nm23-M5|NM23H5|NME/NM23 family member 5|Non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase)|Nonmetastatic cells 5, protein expressed in|Nonmetastatic protein 23, homolog 5|Nucleoside diphosphate kinase homolog 5|Radial spoke 23 homolog|RSPH23|Testis specific nm23 homolog|Testis-specific nm23 homolog
- Inhibitor of p53 induced apoptosis beta|Inhibitor of p53-induced apoptosis-beta|IPIA beta|IPIA-beta|NDK-H 5|NDK5_HUMAN|NDP kinase homolog 5|NM23 H5|nm23-H5|Nm23-M5|NM23H5|NME/NM23 family member 5|Non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase)|Nonmetastatic cells 5, protein expressed in|Nonmetastatic protein 23, homolog 5|Nucleoside diphosphate kinase homolog 5|Radial spoke 23 homolog|RSPH23|Testis specific nm23 homolog|Testis-specific nm23 homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P56597
- P56597
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nucleoside diphosphate kinase homolog 5 protein (1-212AA)
- Nucleoside diphosphate kinase homolog 5 protein (1-212AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together