Order Entry
Canada
ContactUsLinkComponent
Anti-FOXG1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-FOXG1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-FOXG1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77855-549
  • 77737-422
  • Antigen symbol:
  • FOXG1
  • FOXG1
  • Antigen synonyms:
  • BF-1|BF-2|BF1|BF2|Brain factor 1|Brain factor 2|FHKL|FKH2|FKHL1|FKHL2|FKHL3|FKHL4|Forkhead box protein G1|Forkhead box protein G1A|Forkhead box protein G1B|Forkhead box protein G1C|Forkhead like 1|Forkhead like 2|Forkhead like 3|Forkhead like 4|Forkhead-related protein FKHL1|Forkhead-related protein FKHL2|Forkhead-related protein FKHL3|FOXG1_HUMAN|FOXG1A|FOXG1B|FOXG1C|HBF 1|HBF G2|hBF-2|HBF2|HFK1|HFK2|HFK3|KHL2|Oncogene QIN|QIN
  • BF-1|BF-2|BF1|BF2|Brain factor 1|Brain factor 2|FHKL|FKH2|FKHL1|FKHL2|FKHL3|FKHL4|Forkhead box protein G1|Forkhead box protein G1A|Forkhead box protein G1B|Forkhead box protein G1C|Forkhead like 1|Forkhead like 2|Forkhead like 3|Forkhead like 4|Forkhead-related protein FKHL1|Forkhead-related protein FKHL2|Forkhead-related protein FKHL3|FOXG1_HUMAN|FOXG1A|FOXG1B|FOXG1C|HBF 1|HBF G2|hBF-2|HBF2|HFK1|HFK2|HFK3|KHL2|Oncogene QIN|QIN
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P55316
  • P55316
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Forkhead box protein G1 protein (183-292AA)
  • Recombinant Human Forkhead box protein G1 protein (183-292AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together